missing translation for 'onlineSavingsMsg'
Learn More

Erlab Filtrete Halo

Product Code. 16373586
Click to view available options
For Use With (Equipment):
Storage Cabinet
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16373586

Brand: Erlab I1040000001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Filter for your air purifier station.

  • Filter for your air purifier station Halo
  • Dimensions: 190 x 190 mm

Specifications

For Use With (Equipment) Storage Cabinet
Product Type Prefilter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.