Learn More
Witeg™ Filtration Flask
Description
- With PPN plastic hose connection and plastic tubature
- With safety coating for extra protection and durability
- Made of borosilicate glass
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Filtration, vacuum applications |
| Product Type | Filtration flask |
| Shape | Erlenmeyer |
| Bottom Shape | Flat |