Learn More
Macherey-Nagel™ Pleated Filter MN 615 1/4
Use for chemical analysis in waste water treatment. Pleated Filter MN 615 1/4 is perfect for determination of dry matter.
Brand:  Macherey-Nagel™ 531018
Additional Details : Weight : 0.20700kg
 View more versions of this product
Product Code. 10342811
  • 42.05 € / Pack of 100
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Chemical analysis in a waste water treatment plant
  • Ideally suited to be used for analytical procedures 
  • Reproducible results
  • Norm conforming filtration
  • Quality "Made in Germany"
  • Determination of dry matter - filtration for all N-parameters
Specifications
Specifications
Folded
Pleated Filter
185 mm
MN 615 1/4
185 mm