Learn More
Robu™ Filter Funnel
Robu™ Filter Funnels are designed to be use with a Buchner funnel.
55.20 € - 443.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10194121
|
Robu™
21120 |
125 mL |
55.20 €
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
10225841
|
Robu™
21114 |
1000 mL |
195.00 €
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
10614302
|
Robu™
21403 |
4000 mL |
443.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
- Can withstand acids, alkalis, and solvents and even temperatures of up to 540°C
- Filter sucking
- Form according to Büchner
Specifications
Borosilicate Glass | |
Filter Funnel |