Learn More
Cytiva Filter
204.00 € - 1020.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11999217
|
Cytiva
10409872 |
5 cm dia. |
204.00 €
Pack of 100 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
13474547
|
Cytiva
10549778 |
150 x 165 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12425232
|
Cytiva
10381196 |
40 x 330 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12465322
|
Cytiva
2200-654 |
40 mm x 50 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||