Learn More
b.safe Filling Tubes
Product Code. 17191196
Change view
Click to view available options
Quantity:
5/Cs.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17191196 5/Cs. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17191196 Supplier b.safe Supplier No. R43201
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These are Filling tubes made of static dissipative PE-EX material. They are designed to be used with b.safe funnels 180 and have a usable length of 97 mm.
  • Filling tube segments
  • Can be securely connected via clip-in noses to extend as required
  • Each segment has 4 oblong openings on the side for even drainage
Specifications
Length (Metric) 97 mm
For Use With (Equipment) Funnels
Material Expanded Polyethylene
Type Filling Tube
Quantity 5/Cs.
Autoclavable Autoclavable