Learn More
b.safe Filling Tubes
Description
- Filling tube segments
- Can be securely connected via clip-in noses to extend as required
- Each segment has 4 oblong openings on the side for even drainage
Specifications
Specifications
| Length (Metric) | 97 mm |
| For Use With (Equipment) | Funnels |
| Material | Expanded Polyethylene |
| Type | Filling Tube |
| Quantity | 5/Cs. |
| Autoclavable | Autoclavable |