missing translation for 'onlineSavingsMsg'
Learn More

FICZ, MedChemExpress

Product Code. 16415178
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16415178

Brand: Medchem Express HY12451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM.

FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM.

Specifications

Chemical Name or Material FICZ
Physical Form Solid
CAS 172922-91-7
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H12N2O
Synonym 6-Formylindolo[3,2-b]carbazole
Solubility Information DMSO : 10 mg/mL (35.17 mM; Need ultrasonic and warming)
SMILES O=CC1=C2C(NC3=C2C=CC=C3)=CC4=C1NC5=C4C=CC=C5
Molecular Weight (g/mol) 284.31
Formula Weight 284.31
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.