Learn More
FICZ, MedChemExpress
Product Code. 16453306
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16453306
Brand: Medchem Express HY12451
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM.
FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM.
Specifications
Chemical Name or Material FICZ
Physical Form Solid
CAS 172922-91-7
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C19H12N2O
Synonym 6-Formylindolo[3,2-b]carbazole
Solubility Information DMSO : 10 mg/mL (35.17 mM; Need ultrasonic and warming)
SMILES O=CC1=C2C(NC3=C2C=CC=C3)=CC4=C1NC5=C4C=CC=C5
Molecular Weight (g/mol) 284.31
Formula Weight 284.31
Percent Purity 95.0%
Grade Research