missing translation for 'onlineSavingsMsg'
Learn More

FICD Mouse anti-Human, Clone: OTI2A11, TrueMAB™, Origene

Product Code. 17521613
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17521613 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17521613 Supplier Origene Technologies Supplier No. TA802841S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FICD Monoclonal antibody specifically detects FICD in Human samples. It is validated for Western Blot

Specifications

Antigen FICD
Applications Western Blot
Classification Monoclonal
Clone OTI2A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ficd
Gene Accession No. Q9BVA6
Gene Alias adenosine monophosphate-protein transferase FICD; AMPylator FICD; D5Ertd40e; De-AMPylase FICD; FIC domain containing; FIC domain protein adenylyltransferase; FIC domain-containing protein; fic S-phase protein cell division homolog; FICD; HIP13; HIP-13; huntingtin interacting protein 13; huntingtin interacting protein E; huntingtin interactor protein E; huntingtin yeast partner E; Huntingtin-interacting protein 13; huntingtin-interacting protein E; Hype; MGC5623; Protein adenylyltransferase FICD; protein adenylyltransferase FICD; adenosine monophosphate-protein transferase FICD; UNQ3041; UNQ3041/PRO9857
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 61-265 of human FICD produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11153
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.