Learn More
Invitrogen™ Fibulin 2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA597735
Product Code. 16570562
  • 464.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Fibulin 2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibulin 2
Polyclonal
Unconjugated
FBLN2
5730577E14Rik; FBLN2; FIBL2; FIBL-2; fibulin 2; Fibulin2; fibulin-2
Rabbit
Protein G
RUO
2199
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
3.355 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
P98095
FBLN2
Recombinant Human Fibulin-2 protein (301-440AA).