Learn More
Thermo Scientific Chemicals Fibronectin-Binding Protein
Product Code. 15498449
Change view
Click to view available options
Quantity:
0.5mg
1mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498449 0.5mg 0.5mg
15408459 1mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15498449 Supplier Thermo Scientific Chemicals Supplier No. J66903.!D
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
missing translation for 'validMainframeMsg'
For research use
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized powder
Quantity 0.5mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Fibronectin-Binding Protein
Recombinant Native
Form Solid