Learn More
Lonza™ Fibroblast Growth Medium
Specifications
Specifications
| Product Type | Growth Medium |
| Quantity | 1 Kit |
| Content And Storage | Kit which contains a 500mL bottle of FBMTM, (CC-3131) and FGMTM-3 singlequotsTM (CC-4525-supplements and growth factors (hFGF-B, insulin, FBS and gentamicin/ amphotericin-B) |
Safety and Handling
Product Identifier
- FGM-3 Bullet Kit
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: Gentamicin, sulfate(salt)