missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Fibrinogen-Binding Inhibitor Peptide
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Inhibits the binding of fibrinogen and fibronectin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | Fibrinogen γ-chain dodecapeptide |
| Format | Solid |
| For Use With (Application) | Inhibits the binding of fibrinogen and fibronectin |
| Molecular Weight (g/mol) | 1189.30 |
| Product Type | Fibrinogen-Binding Inhibitor Peptide |
| Quantity | 5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | H-His-His-Leu-Gly-Gly-Ala-Lys-Gln-Ala-Gly-Asp-Val-OH |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction