missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FHL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529907
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, mouse brain. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. This protein is thought to have a role in the assembly of extracellular membranes. Also, this gene is down-regulated during transformation of normal myoblasts to rhabdomyosarcoma cells and the encoded protein may function as a link between presenilin-2 and an intracellular signaling pathway. Multiple alternatively spliced variants encoding different isoforms have been identified.
Specifications
| FHL2 | |
| Polyclonal | |
| Unconjugated | |
| Fhl2 | |
| AAG11; aging-associated gene 11; down-regulated in rhabdomyosarcoma LIM protein; DRAL; Fhl2; FHL-2; four and a half LIM domains 2; four and a half LIM domains protein 2; LIM domain protein DRAL; OTTHUMP00000203164; OTTHUMP00000203168; OTTHUMP00000203169; skeletal muscle LIM-protein 3; SLIM 3; SLIM3; SLIM-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14200, 2274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O70433, Q14192 | |
| Fhl2 | |
| Recombinant protein encompassing a sequence within the center region of human FHL2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction