missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1OP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16478
This item is not returnable.
View return policy
Description
FGFR1OP Polyclonal specifically detects FGFR1OP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FGFR1OP | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| FGFR1 oncogene partner, FOPfibroblast growth factor receptor 1 oncogene partner | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cell Cycle and Replication | |
| 11116 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| O95684 | |
| CEP43 | |
| Recombinant protein encompassing a sequence within the center region of human FGFR1OP. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction