missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FGF-Basic Full Length Recombinant Human Protein

Product Code. 11390832
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390832 100 μg Each
11300842 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390832 Supplier Gibco™ Supplier No. CTP0261

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Human FGF-basic (full length)

Cell Therapy Systems CTS™ Recombinant Human FGF-basic (full length) is a bioactive protein intended for use in cell culture applications, including pre-clinical and clinical cell therapy studies. FGF-basic (FGFb), is involved in a number of biological processes including embryonic development and differentiation, and neuron differentiation, survival, and regeneration. FGFb is also involved in the proliferation of cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin. The lyophilized, E. coli-expressed FGF-basic (full length) CTS™ Recombinant Human is verified for activity through dose-dependent proliferation of BALB/3T3 cells.

  • Qualified—helps reduce your burden in qualifying reagents for regulatory submission in cell therapy applications
  • Highly Pure—minimizes interference from other proteins or contaminants
  • Active—get more results with less protein

Simplified Transition into Clinical Research

GIBCO™ Cell Therapy Systems (CTS™) products are of high quality and supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

High Purity

To help verify that GIBCO™ CTS™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO™ CTS™ recombinant growth factor is verified by SDS-PAGE to be over 95% pure and to have endotoxin levels below 0.1 ng/μg.

High Biological Activity

All GIBCO™ CTS™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our FGF-basic (full length) CTS™ Recombinant Human growth factor is determined by the dose-dependent proliferation of BALB/3T3 cells to have an ED50 range of 0.1–1.0 ng/mL (specific activity: 1.0 × 107–1.0 × 106 units/mg). Optimal concentration for individual applications should be determined by a dose response assay.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 17.2 kDa
Purification Method Sequential Chromatography
Quantity 100 μg
Gene Alias FGF
Protein Length 155 aa
Species Human
Recombinant Recombinant
Content And Storage 2°C to 8°C
Classification Carrier-Free
Activity 0.1 ng/ml
Bioassay Proliferation of BALB⁄3T3 cells
Endotoxin Level <0.1 ng/μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Growth Factors & Receptors
Protein Form Full Length, Recombinant, Ligand
Form Lyophilized
Protein Subtype FGFs (Fibroblast Growth Factors)
Research Category Clinical Research, Oncology, Angiogenesis, Stem Cell Research, Microbiology, Differentiation, Neurobiology
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.