Learn More
Hach Lange™ FerroZine™ Iron Reagent Solution Pillows
Product Code. 10588912
Click to view available options
Quantity:
25 mL
Unit Size:
Pack of 50
Product Code. 10588912
Brand: Hach Lange™ 230166
Please to purchase this item. Need a web account? Register with us today!
Hach Lange FerroZine Iron Reagent Solution Pillows, Quantity: 25mL, Method: FerroZine
Specifications
Quantity 25 mL
Test Method FerroZine