missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferredoxin Reductase Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Ferredoxin Reductase Polyclonal antibody specifically detects Ferredoxin Reductase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Ferredoxin Reductase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Adrenodoxin reductase, ADXREC 1.18.1.2, ferredoxin reductaseAR, Ferredoxin--NADP(+) reductase, NADPH:adrenodoxin oxidoreductase, mitochondrial |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 252-491 of human Ferredoxin Reductase (NP_077728.3).,, Sequence:, PILDPVDFLGLQDKIKEVPRPRKRLTELLLRTATEKPGPAEAARQASASRAWGLRFFRSPQQVLPSPDGRRAAGVRLAVTRLEGVDEATRAVPTGDMEDLPCGLVLSSIGYKSRPVDPSVPFDSKLGVIPNVEGRVMDVPGLYCSGWVKRGPTGVIATTMTDSFLTGQMLLQDLKAGLLPSGPRPGYAAIQALLSSRGVRPVSFSDWEKLDAEEVARGQGTGKPREKLVDPQEMLRLLGH |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?