Learn More
Fenthion Sulfoxide-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Fenthion-sulfoxide D6 (O,O-dimethyl D6) |
| CAS | 2733270-79-4 |
| Molecular Formula | C10 2H6 H9 O4 P S2 |
| Quantity | 10 mg |
| Synonym | Fenthion Sulfoxide D6 (O,O-dimethyl D6) |
| InChI Formula | InChI=1S/C10H15O4PS2/c1-8-7-9(5-6-10(8)17(4)11)14-15(16,12-2)13-3/h5-7H,1-4H3/i2D3,3D3 |
| SMILES | [2H]C([2H])([2H])OP(=S)(Oc1ccc(c(C)c1)S(=O)C)OC([2H])([2H])[2H] |
| IUPAC Name | (3-methyl-4-methylsulfinylphenoxy)-sulfanylidene-bis(trideuteriomethoxy)-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 300.36 |
| Formula Weight | 300.0526 |