missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ Fenoprofen calcium

Product Code. 18749456
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18749456 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18749456 Supplier Tocris™ Supplier No. 6926/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Positive allosteric modulator of MC3 receptors; exhibits biased signaling

Fenoprofen calcium is a melanocortin 3 (MC3) receptor positive allosteric modulator (PAM; allosteric enhancer) (pEC50 = 8.69); decreases the EC50 of α-MSH at MC3R. Stimulates ERK1/2 activation at MC3-5R, with highest ERK1/2 activation observed at 3 and 10 μM for MC3R, 1 μM for MC4R, and 3 μM for MC5R. Exhibits biased signaling at MC3-5R, selectively activating ERK1/2 cascade but not canonical cAMP signaling. Antiarthritic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material α-Methyl-3-phenoxybenzeneacetic acid hemicalcium salt
CAS 34597-40-5
Quantity 50 mg
Molecular Formula C15H13O3.0.5Ca
Formula Weight 261.31
Product Type Positive Allosteric Modulator
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.