Learn More
Sartorius Female Luer Fittings
Product Code. 17889366 Shop All Sartorius Products
Click to view available options
Material:
Tefzel
Unit Size:
1 set
Product Code. 17889366
Brand: Sartorius 17002140
Only null left
Add to basket
Add to basket
Adapters to convert the Luer inlet and outlet connectors on any Sartobind™ Lab unit to the M6 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with M6 male connectors.
  • Adapters to convert the Luer inlet and outlet connectors on any Sartobind™ Lab unit to the M6 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with M6 male connectors.
  • 2 x M6 female to female Luer and 2 x M6 female to male Luer adapters
  • Black Tefzel
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Color Black
Material Tefzel
Pressure 100 psi