Learn More
Sartorius Female Luer Fittings
Shop All Sartorius Products
Click to view available options
Material:
Tefzel
Unit Size:
1 set
Description
- Adapters to convert the Luer inlet and outlet connectors on any Sartobind™ Lab unit to the M6 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with M6 male connectors.
- 2 x M6 female to female Luer and 2 x M6 female to male Luer adapters
- Black Tefzel
Specifications
Specifications
| Type | Adapter |
| Color | Black |
| Material | Tefzel |
| Pressure | 100 psi |