missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Felvizumab Recombinant Human Monoclonal Antibody (RSHZ19 (Felvizumab))
GREENER_CHOICE

Product Code. 17991051
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
Product Code. Quantity unitSize
17991051 200 μg 200µg
1 options
This item is not returnable. View return policy

Product Code. 17991051

Brand: Invitrogen™ MA548053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Specificity: This antibody is specific for the an epitope located between amino acids 417-432 of conserved fusion (F) protein of RSV.

Felvizumab is a biosimilar that targets RSV. Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Felvizumab
Applications ELISA, Functional Assay
Classification Recombinant Monoclonal
Clone RSHZ19 (Felvizumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias SB 209763
Host Species Human
Immunogen This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.