Learn More
Felodipine 3,5-Dimethyl Ester, TRC
Product Code. 30406409
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406409 100 mg
30406291 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30406409 Supplier Toronto Research Chemicals Supplier No. TRCF232385100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 91189-59-2
Molecular Formula C17 H17 Cl2 N O4
Molecular Weight (g/mol) 370.23
Synonym Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Felodipine Imp. B (EP)
IUPAC Name dimethyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)OC)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C17H17Cl2NO4/c1-8-12(16(21)23-3)14(10-6-5-7-11(18)15(10)19)13(9(2)20-8)17(22)24-4/h5-7,14,20H,1-4H3
Formula Weight 369.0535
Percent Purity >95%
Chemical Name or Material Dimethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Grade HPLC