Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, stainless steel, 16ga x 76mm, straight, sterile
Click to view available options
Product Type:
Feeding Tube
Description
These traditional reusable gavage needles feature stainless steel tubes and ball-tips. They are sold straight but are easily bent to your desired curve for optimal placement. Available in seven sizes for mice and rats.
- Stainless steel feeding tubes, 16ga x 76mm
- For rodent oral gavage
- 2.8mm round bulb
- Bendable
- Supplied as sterile pouch of 6 tubes
Specifications
Specifications
| Product Type | Feeding Tube |