Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, polypropylene, 18ga x 75mm, sterile
Product Code. 15135024
Encompass
Change view
Click to view available options
Product Type:
Feeding Tube
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15135024 Feeding Tube Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15135024 Supplier Instech Laboratories™ Supplier No. FTP187550
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Instech's polypropylene gavage tubes are flexible to reduce trauma, translucent to visualise delivery, and disposable to eliminate the cost of cleaning and the chance of compound crossover. Flexible tubes are a recommended animal welfare refinement over rigid metal tubes. The tube and luer hub are made of the same material used to make plastic syringes. The soft, bulb-shaped tip is designed to reduce the chances of inadvertent placement in the trachea and perforation of the oesophagus. Available in 11 sizes for rodents ranging from mouse pups to rats to rabbits.
  • Plastic feeding tubes, 18ga x 75mm
  • For rodent oral gavage
  • Designed to minimise trauma to oesophagus
  • Polypropylene female luer (pink) and flexible polypropylene tube with soft elastomer tip
  • Disposable
  • Supplied as 10 sterile pouches of 5 tubes
Specifications
Product Type Feeding Tube