Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, polypropylene, 18ga x 38mm, sterile
Product Code. 15300340
Encompass
Click to view available options
Product Type:
Feeding Tube
This item is not returnable. View return policy
Product Code. 15300340
Brand: Instech Laboratories™ FTP1838
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Instech's polypropylene gavage tubes are flexible to reduce trauma, translucent to visualise delivery, and disposable to eliminate the cost of cleaning and the chance of compound crossover. Flexible tubes are a recommended animal welfare refinement over rigid metal tubes. The tube and luer hub are made of the same material used to make plastic syringes. The soft, bulb-shaped tip is designed to reduce the chances of inadvertent placement in the trachea and perforation of the oesophagus. Available in 11 sizes for rodents ranging from mouse pups to rats to rabbits.
  • Plastic feeding tubes, 18ga x 38mm
  • For rodent oral gavage
  • Designed to minimise trauma to oesophagus
  • Polypropylene female luer (pink) and flexible polypropylene tube with soft elastomer tip
  • Disposable
  • Supplied as 50 sterile pouches of 5 tubes
Specifications
Product Type Feeding Tube