Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, polypropylene, 13ga x 150mm, sterile
Description
Instech's polypropylene gavage tubes are flexible to reduce trauma, translucent to visualise delivery, and disposable to eliminate the cost of cleaning and the chance of compound crossover. Flexible tubes are a recommended animal welfare refinement over rigid metal tubes. The tube and luer hub are made of the same material used to make plastic syringes. The soft, bulb-shaped tip is designed to reduce the chances of inadvertent placement in the trachea and perforation of the oesophagus. Available in 11 sizes for rodents ranging from mouse pups to rats to rabbits.
- Plastic feeding tubes, 13ga x 150mm
- For rodent oral gavage
- Designed to minimise trauma to oesophagus
- Polypropylene female luer (purple) and flexible polypropylene tube with soft elastomer tip
- Disposable
- Supplied as 50 sterile pouches of 5 tubes
Specifications
Specifications
| Product Type | Feeding Tube |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.