Learn More
Instech Laboratories™ Feeding tube, rat oral gavage, polyurethane, 13ga x 88mm, sterile
Product Code. 15380340
Encompass
Change view
Click to view available options
Product Type:
Feeding Tube
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15380340 Feeding Tube Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15380340 Supplier Instech Laboratories™ Supplier No. FTPU138850
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
These specialty tubes feature a larger inner diameter for easier delivery of viscous compounds and a straight tip that wipes clean to help in cases where aspiration of compounds would be hazardous to the animal. Availablein two sizes: 13ga for rats and 16ga for mice.
  • Plastic feeding tubes, 13ga x 88mm
  • For rat oral gavage
  • Designed to minimise trauma to oesophagus
  • Polyurethane tube with rounded tip
  • Large internal diameter for viscous compounds
  • Straight tube wipes clean to prevent aspiration of compounds
  • Disposable
  • Supplied as 10 sterile pouches of 5 tubes
Specifications
Product Type Feeding Tube