Learn More
FDI-6, MedChemExpress
Product Code. 16474688
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474688 10 mg 10mg
16427407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16474688 Supplier Medchem Express Supplier No. HY112721
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress FDI-6 is an inhibitor of FOXM1. FDI-6 binds directly to FOXM1 protein, to displace FOXM1 from genomic targets in MCF-7 breast cancer cells, and induce concomitant transcriptional down-regulation.
FDI-6 is an inhibitor of FOXM1. FDI-6 binds directly to FOXM1 protein, to displace FOXM1 from genomic targets in MCF-7 breast cancer cells, and induce concomitant transcriptional down-regulation.
Specifications
Chemical Name or Material FDI-6
CAS 313380-27-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H11F4N3OS2
Quantity 10 mg
Solubility Information DMSO : 41.67 mg/mL (95.26 mM; Need ultrasonic)
SMILES O=C(C1=C(N)C2=C(C(F)(F)F)C=C(C3=CC=CS3)N=C2S1)NC4=CC=C(F)C=C4
Molecular Weight (g/mol) 437.43
Formula Weight 437.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death