missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fc OxyBURST™ Green Assay Reagent

Product Code. 10770964
Click to view available options
Quantity:
500 μL, 25 reactions
Unit Size:
Each
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 10770964

Brand: Invitrogen™ F2902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Fc OxyBURST™ Green Assay Reagent

The Fc OxyBURST™ Green assay reagent comprises bovine serum albumin (BSA) that has been covalently linked to dichlorodihydrofluorescein (H2DCF) and then complexed with a purified rabbit polyclonal anti-BSA antibody. When this immune complex binds to Fc receptors, the nonfluorescent H2DCF molecules are internalized within the phagovacuoloe and subsequently oxidized to green-fluorescent 2''7'-dichlorofluorescein.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 3 mg⁄ml
Content And Storage Contains 1 vial of Fc OxyBURST™ assay green reagent (suspension in PBS containing sodium azide). Store in refrigerator (2–8°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Assay Reagent
Dye Type Free Radical Sensor
Emission 488/523
Form Solution
Format Tube(s), Slide(s)
Product Line OXYBURST
Quantity 500 μL, 25 reactions
Shipping Condition Wet Ice
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.