Learn More
FBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16021-1-AP-150UL
This item is not returnable.
View return policy
Description
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13.1).Specifications
| FBL | |
| Polyclonal | |
| Unconjugated | |
| FBL | |
| FBL, FIB, FIB1, fibrillarin, FLRN, RNU3IP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14113, 2091, 292747 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22087, P22509, P35550 | |
| FBL | |
| FBL Fusion Protein Ag8841 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |