missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fatty Acid Synthase/FASN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49213
This item is not returnable.
View return policy
Description
Fatty Acid Synthase/FASN Polyclonal antibody specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Fatty Acid Synthase/FASN | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MPFRGYAVLGGERGGPEVQQVPAGERPLWFICSGMGTQWRGMGLSLMRLDRFRDSILRSDEAVKPFGLKVSQLLLSTDESTFDDIVHSFVSLTAIQIGLIDLLSCMGLRPDGIVG | |
| 0.1 mL | |
| Apoptosis, Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology | |
| 2194 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction