missing translation for 'onlineSavingsMsg'
Learn More

FATE1 Monoclonal Antibody (OTI3D7), TrueMAB™, OriGene

Product Code. 17543332
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17543332 30 μL 30µL
17523332 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17543332 Supplier Origene Technologies Supplier No. TA505182S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FATE1 Monoclonal antibody specifically detects FATE1 in Human samples. It is validated for Western Blot

Specifications

Antigen FATE1
Applications Western Blot
Classification Monoclonal
Clone OTI3D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FATE1
Gene Accession No. Q969F0
Gene Alias 9230118O15Rik; BJ-HCC-2 antigen; cancer/testis antigen 43; CT43; EGK_21049; FATE; FATE1; fetal and adult testis expressed 1; fetal and adult testis expressed transcript protein; fetal and adult testis-expressed transcript protein; Tumor antigen BJ-HCC-2
Host Species Mouse
Immunogen Full length human recombinant protein of human FATE1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89885
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.