missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FASTKD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16617664
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16617664 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16617664 Supplier Invitrogen™ Supplier No. PA5101182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAKD1.

As a member of the FASTKD family, FASTKD1 localizes to the mitochondria to modulate their energy balance, especially under conditions of stress. Though ubiquitously expressed in all tissues, FASTKD1 appears more abundantly in skeletal muscle, heart muscle, and other tissues enriched in mitochondria. FASTKD1 has been validated as an RNA-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASTKD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FASTKD1
Gene Accession No. Q53R41, Q6DI86
Gene Alias 5330408N05Rik; FAST kinase domain-containing protein 1; FAST kinase domain-containing protein 1, mitochondrial; FAST kinase domains 1; Fastkd1; KIAA1800; mKIAA1800; RGD1563531
Gene Symbols FASTKD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FASTKD1(Accession Q53R41), corresponding to amino acid residues V560-P610.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320720, 79675
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.