Learn More
Fasiglifam, MedChemExpress
Product Code. 16484996
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484996 100 mg 100mg
16443998 10 mg 10mg
16466707 5 mg 5mg
16420116 2 mg 2mg
16480986 200 mg 200mg
16490347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16484996 Supplier Medchem Express Supplier No. HY10480
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Fasiglifam (TAK-875) is a potent, selective and orally bioavailable GPR40 agonist with EC50 of 72 nM.
Fasiglifam (TAK-875) is a potent, selective and orally bioavailable GPR40 agonist with EC50 of 72 nM.
Specifications
Chemical Name or Material Fasiglifam
CAS 1000413-72-8
Color Red
Purity Grade Notes Research
Molecular Formula C29H32O7S
Quantity 100 mg
Synonym TAK-875
Solubility Information DMSO : ≥ 128 mg/mL (243.98 mM)
SMILES CC1=C(C(C)=CC(OCCCS(C)(=O)=O)=C1)C2=CC(COC3=CC=C4[C@@H](COC4=C3)CC(O)=O)=CC=C2
Molecular Weight (g/mol) 524.63
Formula Weight 524.63
Percent Purity 95.05%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism