missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM83G Polyclonal Antibody
GREENER_CHOICE

Product Code. 15772701 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15772701 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15772701 Supplier Invitrogen™ Supplier No. PA554775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RLLPDPGSPR LAQNARPMTD GRATEEHPSP FGIPYSKLSQ SKHLKARTGG SQWASSDSKR RAQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

May regulate the bone morphogenetic proteins (BMP) pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83G
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM83G
Gene Accession No. A6ND36
Gene Alias FAM83G; family with sequence similarity 83 member G; family with sequence similarity 83, member G; PAWS1; Protein associated with SMAD1; protein FAM83G
Gene Symbols FAM83G
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM83G. Recombinant protein control fragment (Product #RP-93532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644815
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.