Learn More
FAM40A Monoclonal Antibody (OTI7B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502310
Description
Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the cortical actin filament dynamics and cell shape.Specifications
| FAM40A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5VSL9 | |
| Strip1 | |
| Full length human recombit protein of human FAM40A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7B8 | |
| Unconjugated | |
| STRIP1 | |
| FAM40A, FAR11A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 362012, 490117, 85369 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.