missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM3D Monoclonal Antibody (K29027_4E9), Invitrogen™

Product Code. 30207683
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30207683 200 μL 200µg
30212336 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30207683 Supplier Invitrogen™ Supplier No. MA556185

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: YMSFSMKTIR LPRWLAASPT KEIQVKKYKC GLIKPCPANY FAFKICSGAA NVVGPTMCFE DRMIMSPVKN NVGRGLNIAL VNGTTGAVLG QKAFDMYSGD VMHLVKFLKE IPGGALVLVA SYDDPGTKMN DESRKLFSDL GSSYAKQLGF RDSWVFIGAK DLRGKSPFEQ FLKNSPDTNK YEGWPELLEM EGCMPPKPF

FAM3D (FAM3 Metabolism Regulating Signaling Molecule D) is a Protein Coding gene. Diseases associated with FAM3D include Deafness, Autosomal Recessive 17 and Scheuermann Disease. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is FAM3C. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM3D
Applications Western Blot
Classification Monoclonal
Clone K29027_4E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Fam3d
Gene Accession No. Q96BQ1
Gene Alias 2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; Fam3d; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; oncoprotein-induced protein 1; Protein EF-7; protein FAM3D; UNQ567/PRO1130
Gene Symbols Fam3d
Host Species Mouse
Immunogen Recombinant human FAM3D (immunogen range is 26-224).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.