missing translation for 'onlineSavingsMsg'
Learn More

FAM127A Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16866603 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16866603 20 μL 20µL
16856603 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16866603 Supplier Proteintech Supplier No. 103071AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody specifically recognizes the O15255.

RTL8C (Retrotransposon Gag Like 8C) is a Protein Coding gene. An important paralog of this gene is RTL8A.

Specifications

Antigen FAM127A
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RTL8C
Gene Accession No. O15255
Gene Alias CAAX box protein 1, CXX1, FAM127A, Mar8, MAR8C, Mart8, MART8C, Protein FAM127A
Gene Symbols RTL8C
Host Species Rabbit
Immunogen FAM127A Fusion Protein Ag0298
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8933
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.