missing translation for 'onlineSavingsMsg'
Learn More

Fam Benzine, Honeywell

Product Code. p-7073677
Click to view available options
Quantity:
2.5 L
25 L
Unit Size:
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15642670

Brand: Honeywell Riedel-de Haën 245382.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

DIN 51635, bp 65-95 °C

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.690 to 0.705g/mL at 15°C
Flash Point −26°C (−14.8°F)
Quantity 2.5 L
UN Number 1268
Chemical Name or Material Fam Benzine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.