Learn More
FAHD2A Monoclonal Antibody (OTI7C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500946S
Description
FAHD2A Monoclonal antibody specifically detects FAHD2A in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FAHD2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96GK7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51011 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI7C9 | |
| Unconjugated | |
| FAHD2A | |
| 1500003K14Rik; B430104H02Rik; CGI-105; CGI-105 protein; Fahd2; FAHD2A; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain containing 2A; fumarylacetoacetate hydrolase domain-containing protein 2; Fumarylacetoacetate hydrolase domain-containing protein 2A; LOC100062565; LOC100731203; RGD1563674 | |
| Full length human recombinant protein of human FAHD2A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.