Learn More
FAHD2A Monoclonal Antibody (OTI7C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500946
Description
May have hydrolase activity.Specifications
| FAHD2A | |
| Monoclonal | |
| Unconjugated | |
| FAHD2A | |
| CGI-105 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51011 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI7C9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96GK7 | |
| FAHD2A | |
| Full length human recombit protein of human FAHD2A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.