Learn More
FABP2/I-FABP Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1486-SP
This item is not returnable.
View return policy
Description
FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Product Title