Learn More
FABP2/I-FABP Goat anti-Rat, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF1486-SP
Encompass
Product Code. 15760904
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.