Learn More
Fabomotizole hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Fabomotizole hydrochloride (CM346 hydrochloride) is an anxiolytic drug. Fabomotizole hydrochloride produces anxiolytic and neuroprotective effects without any muscle relaxant actions.
Specifications
Specifications
| Chemical Name or Material | Fabomotizole hydrochloride |
| CAS | 173352-39-1 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H22ClN3O2S |
| Quantity | 5 mg |
| Synonym | CM346 hydrochloride |
| Solubility Information | H2O : 100 mg/mL (290.81 mM; Need ultrasonic) ∣DMSO : ≥ 44 mg/mL (127.96 mM) |
| SMILES | CCOC1=CC=C2C(N=C(SCCN3CCOCC3)N2)=C1.Cl |
| Molecular Weight (g/mol) | 343.87 |
| Show More |