missing translation for 'onlineSavingsMsg'
Learn More

FAAH-IN-2, MedChemExpress

Product Code. p-200096179
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16470688

Brand: Medchem Express HY79511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FAAH-IN-2 (O-Desmorpholinopropyl Gefitinib) is a potent FAAH(fatty acid amide hydrolase) inhibitor extracted from Patent WO/2008/100977A2.

FAAH-IN-2 (O-Desmorpholinopropyl Gefitinib) is a potent FAAH(fatty acid amide hydrolase) inhibitor extracted from Patent WO/2008/100977A2.

Specifications

Chemical Name or Material FAAH-IN-2
CAS 184475-71-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H11ClFN3O2
Quantity 5 g
Synonym O-Desmorpholinopropyl Gefitinib
Solubility Information DMSO : 20.83 mg/mL (65.15 mM; Need ultrasonic)
SMILES OC1=CC2=C(NC3=CC=C(F)C(Cl)=C3)N=CN=C2C=C1OC
Molecular Weight (g/mol) 319.72
Formula Weight 319.72
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.