missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Rat, anti-Mouse, Biotin, Clone: BM8, STEMCELL Technologies
Description
- The BM8 antibody reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ∼160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors
- F4/80 is considered a marker of choice for the identification of mature tissue macrophages, being broadly but variably expressed by this cell type in the liver (Kupffer cells), skin (Langerhans cells), bone marrow stroma, pancreas, thymus, spleen (red pulp), lung, and other tissues
- It is also expressed by circulating monocytes, eosinophils and a subset of dendritic cells
- F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770)
Specifications
Specifications
| Antigen | F4/80 |
| Applications | Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | BM8 |
| Conjugate | Biotin |
| Host Species | Rat |
| Quantity | 50 μg |
| Primary or Secondary | Primary |
| Target Species | Human, Mouse |
| Isotype | IgG2a κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?