missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 546
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ A11071
This item is not returnable.
View return policy
Description
Product will be shipped at Room Temperature.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| Alexa Fluor 546 | |
| Goat | |
| Purified | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 5mM sodium azide; pH 7.5 | |
| Gamma Immunoglobins Heavy and Light chains. | |
| 500 μg | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction