Learn More
F 16915, MedChemExpress
Product Code. 16477948
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477948 10 mg 10mg
16431889 1 mg 1mg
16410418 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16477948 Supplier Medchem Express Supplier No. HY19886
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
MedChemExpress F 16915, a Docosahexaenoic Acid (DHA, HY-B2167) derivative, is a potent pro-drug of DHA. F 16915 can prevent heart failure-induced atrial fibrillation.
F 16915, a Docosahexaenoic Acid (DHA, HY-B2167) derivative, is a potent pro-drug of DHA. F 16915 can prevent heart failure-induced atrial fibrillation.
Specifications
Chemical Name or Material F 16915
Physical Form Oil
CAS 92510-91-3
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H37NO2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (238.32 mM)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(OCC1=CC=CN=C1)=O
Molecular Weight (g/mol) 419.6
Formula Weight 419.6
Percent Purity 99.39%
Grade Research