Learn More
Thermo Scientific™ EZ-Link™ TFP-PEG3-Biotin
Product Code. 10628474 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10628474 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10628474 Supplier Thermo Scientific™ Supplier No. 21219
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Biotinylate primary amines (e.g., protein lysines) and secondary amines such as histidine and tryptophan with this tetrafluorophenyl-ester biotinylation reagent.
Thermo Scientific™ EZ-Link TFP-PEG3-Biotin is a tetrafluorophenyl-ester biotinylatoin reagent that enables labeling at primary amines found on lysine residues and the N-terminus and secondary amines such as histidine and tryptophan.
Highlights:
  • Tetrafluorophenyl (TFP) ester reacts with primary and secondary amines at pH 7-9
  • Long-chain, water-soluble, polyethylene glycol (PEG) spacer arm
  • PEG spacer arm transfers hydrophilicity to final conjugate
  • Creates an amide bond
TRUSTED_SUSTAINABILITY
Specifications
Cleavable No
Description EZ-Link TFP-PEG3-Biotin
Formulation Tetrafluorophenyl ester of biotin with a triethyleneglycol spacer
Molecular Weight (g/mol) 694.74
Membrane Permeable No
PEGylated Yes
Spacer Arm Length 32.6
Quantity 50 mg
Sufficient For Approx. 25 uses to label a total of 480mg of IgG using typical conditions
Chemical Reactivity Amine
Solubility Water