Learn More
Thermo Scientific™ EZ-Link™ Alkoxyamine-PEG12-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with a very long (55.4A), pegylated biotinylation reagent containing a 12-unit PEG spacer for length and solubility.
Brand: Thermo Scientific™ 26139
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific EZ-Link Alkoxyamine-PEG12-Biotin is a biotinylation reagent containing a very long, 12-unit polyethylene glycol (PEG) spacer for biotinylating macromolecules at carbohydrate groups that have been oxidized to form aldehydes.
Alkoxyamine-activated biotinylation reagents such as this one are useful for biotin-labeling glycoproteins and other molecules that have oxidizable polysaccharides groups. EZ-Link Alkoxyamine-PEG-Biotin reagents contain a multi-functional extended spacer arm that is a flexible, non-immunogenic hydrophilic polyethylene glycol (PEG), which imparts water solubility to labeled molecules. Consequently, antibodies labeled with biotin alkoxyamine reagents exhibit less aggregation when stored in solution compared to antibodies labeled with reagents having only hydrocarbon spacers.Highlights:
Biotin – label macromolecules for detection or affinity purification with avidin and streptavidin
Alkoxyamine-activated – reacts with aldehyde groups (e.g., oxidized sugars in glycoproteins) to attach biotin by stable oxime bonds
Polyethylene glycol – PEG groups are flexible, non-immunogenic, hydrophilic, and improve the solubility of labeled antibodies
Long spacer arm – the reach between the point of attachment and the biotin affinity tag is more than 55 angstroms, minimizing steric hindrance for binding
Specifications:
- Chemical formula: C34H66N4O14S
- Molecular weight: 786.97
- Spacer arm: 55.4 angstroms
- Net mass added to target: 768.95
- Form: clear to light amber highly viscous liquid
- Solubility: dissolve in DMSO to make 250mM stock
Specifications
Alkoxyamine-PEG12-Biotin | |
Store at -20°C. Shipped at ambient temperature. | |
Cell-Impermeant | |
Alkoxyamine | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Ketone, Aldehyde | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Extra-long, Pegylated |
For Research Use Only. Not for use in diagnostic procedures.